Eagle CRA-1917 Polyethylene Shelf Tray for Acid Safety Cabinet, 60-gallon
Representative image only.
Eagle Mfr # CRA-1917 – Item # EW-09418-64
Epoxy-coated steel offers protection from chemical attack
  • Shelves direct spills to back and bottom of leakproof sump
  • Trilingual warning label
  • Meets NFPA Code 30 and OSHA requirements
  • FM-Approved
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Gal)0
  • Self-Closingshelf tray only
  • DescriptionPolyethylene Shelf Tray for Acid Safety Cabinet, 60-gallon
Key Features
  • Shelves direct spills to back and bottom of leakproof sump
  • Trilingual warning label
  • Meets NFPA Code 30 and OSHA requirements
  • FM-Approved
More About this Item
These acid corrosive safety cabinets are constructed of a double-walled, 18-gauge steel with 1 1/2" (3.8 cm) insulating barrier, ideal for both flammable and nonflammable acids and corrosive liquids. Includes two vents, each with 2” (5.1 cm) threaded fittings, a fire baffle, and cap.
Cabinets have a corrosion-resistant powder coat finish and feature grounding attachment, 3-point latch system, and 2” (5.1 cm) raised, leakproof door sill. Each HDPE shelf is adjustable on 3” (7.6 cm) centers, supported by 4 brackets, with a 350 lbs (159 kg) safe allowable load. The shelf and sump trays help contain small spills and provide extra corrosion protection. Cabinets are blue with trilingual warning label: CAUTION-CORROSIVES.
Not recommended for use with Sulfuric, Nitric or Hydrochloric acids.