Ebro 1340-5100 TFE 100 Compact Food Thermometer with Remote Probe
Representative image only.
Ebro Mfr # 1340-5100 – Item # EW-91210-35
Ideal for use in the food industry
  • IP55-rated waterproof case
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType T
  • Min Temperature (° F)-58
  • Max Temperature (° F)660
  • Min Temperature (° C)-50
  • Max Temperature (° C)350
  • Resolution0.1°; 1° above 390°F (199°C) and below -76°F (-60°F)
  • IP RatingIP55
  • Display Type3.5-digit LCD
  • Height (in)3 1/2
  • Depth (in)3/4
  • No. of Probe(s) Included1
  • Battery Life100 hours
  • Battery3v Battery (1)
  • DescriptionTFE 100 Compact Food Thermometer with Remote Probe
  • Warranty2 year
Two year warranty
Key Features
  • IP55-rated waterproof case
  • 2 year warranty
More About this Item
These thermometers are constructed of ABS plastic and have an IP55 rating producing a waterproof and shock-resistant product ideal for use in the food industry. The use of an advanced internal microprocessor that updates every five seconds and a highly accurate type T thermocouple produces an accuracy of ±0.8%. The product features a Hold function, °F/°C selectable units, and auto power-off after 15 seconds of inactivity to save on battery power.