Cole-Parmer Economical Infrared Temperature Calibrator, English/French Version
Representative image only.
Cole-Parmer Item # EW-35625-86
Easily calibrate your infrared calibrator
  • 3 year warranty
$4.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionEconomical Infrared Temperature Calibrator, English/French Version
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Simpy aim your IR thermometer at the black area and compare the temperature on the IR thermometer to the green reading on the temperature strip.
Instructions on calibration mat are provided in English and French.
Calibrate temperature range: 26 to 56°F (-3 to 13°C)