Economical Push-Button Temperature Transmitter, Thermocouple
Representative image only.
Item # EW-94771-00
Customize your temperature range with a button press
  • LED indicates programming operations and out-of-range sensor
  • Drift-free digital linearization
  • Fits standard DIN probe head enclosures
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Control Input ThermocoupleJ, K, T
  • Output4 to 20 mA (reversible)
  • Min Temperature (° F)-328
  • Max Temperature (° F)2192
  • Accuracy of Span±0.04%
  • DescriptionEconomical Push-Button Temperature Transmitter, Thermocouple
  • Warranty5 years
5 Year Warranty
Key Features
  • LED indicates programming operations and out-of-range sensor
  • Drift-free digital linearization
  • Fits standard DIN probe head enclosures
  • 5 year warranty
More About this Item
These economical transmitters feature a push-button, customizable temperature range. Simply connect a power supply and a simulator/calibrator to the transmitter, select the desired temperature at 4 or 20 mA, and hold the set button until the LED indicator blinks. Transmitters are factory set at a range of 32 to 212°F (0 to 100°C).
Thermocouple Transmitter is selectable for Type J, K, or T probes. It features galvanic isolation.
RTD Transmitter connects to any standard 3-wire Pt 100 RTD temperature sensor. The RTD model also allows push-button calibration.
Thermistor Transmitter is compatible with 400 series thermistor probes for excellent accuracy over the ambient temperature range.