Edwards C100-09-173 Centering Ring, 100 ISO-K - Nitrile (Vacuum Pump Accessories)
Representative image only.
Edwards Mfr # C100-09-173 – Item # EW-31401-84
Create a tight vacuum seal with these centering rings
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialNitrile
  • Connection TypeVacuum Flange
  • Flange Size (Port 1)100 ISO-K
  • DescriptionCentering Ring, 100 ISO-K - Nitrile (Vacuum Pump Accessories)
More About this Item
Polymer support and nitrile O-rings withstand up to 10–7 Torr (mm Hg). The temperature range of these centering rings is 14 to 212°F (–10 to 100°C).
The centering ring has an elastomer O-ring creating the seal, and inside of this there is a plastic support ring.