Edwards Vacuum C36000100 Vacuum ball valve, NW16 41 to 149 F (5 to 65 C) ambient
Representative image only.
Edwards Vacuum Mfr # C36000100 – Item # EW-79302-63
Excellent in applications where fast action and full bore pumping are needed
  • Rugged design for dirty system use
  • Corrosion resistant — 316L stainless steel with PTFE seats
  • Leak rate better than 1 x 10–6 mbar/sec (8 x 10–7 Torr/sec)
PRODUCT OPTIONS
$309.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortNW16 flange
  • Connection TypeFlanged
  • Inlet PortNW16 flange
  • Max Temperature (° F)149
  • Max Temperature (° C)65
  • DescriptionVacuum ball valve, NW16 41 to 149 F (5 to 65 C) ambient
Key Features
  • Rugged design for dirty system use
  • Corrosion resistant — 316L stainless steel with PTFE seats
  • Leak rate better than 1 x 10–6 mbar/sec (8 x 10–7 Torr/sec)
More About this Item
These Edwards Vacuum 316L Stainless Steel (SS) Vacuum Ball Valves are ideal is rugged applications that require corrosion resistance. All valves feature and operating range of 10-6 to 7 bar (8 x 10-7 to 5250 torr) and a temperature range of 41 to 149°F (5 to 65°C).