Cole-Parmer Glass Capillary Tubes, Closed Both Ends, 2.0 mm OD; 1000/PK
Representative image only.
Cole-Parmer Item # EW-03010-68
  • 10 x 100 capillary tubes
  • 2.0 mm OD
  • Previous part number AT4043
PRODUCT OPTIONS
$500.00USD / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Compatible WithMelting point apparatus 03010-15, 03010-41, 03010-51, 03010-59, and 03010-64
  • Diameter (mm)2
  • DescriptionCapillary Tubes; 2.0 mm OD; 1,000/PK
Key Features
  • 10 x 100 capillary tubes
  • 2.0 mm OD
  • Previous part number AT4043
More About this Item
Accessories compatible with digital melting point apparatus.