EMD Millipore 10006-1 Water Test Strips, Nickel, 0 to 500 ppm, 100/Pk
Representative image only.
EMD Millipore Mfr # 1.10006.0001 – Item # EW-99572-07
Use for quick spot checks to ensure water quality
  • Convenient, on-site water testing
SPECS & DESCRIPTION
Specifications & Description
  • TestNickel
  • Measurement Points0, 10, 25, 100, 250, 500 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionWater Test Strips, Nickel, 0 to 500 ppm, 100/Pk
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.