EMD Millipore 10046-1 Total Hardness Test Strips, 90 to 450 ppm
Representative image only.
EMD Millipore Mfr # 1.10046.0001 – Item # UX-99572-11
Use for quick spot checks to ensure water quality
  • Convenient, on-site water testing
$105.25USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • TestHardness
  • Measurement Points90, 180, 270, 360, 450 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionTotal Hardness Test Strips, 90 to 450 ppm
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.