EMD Millipore HYCON® 941225 DG18 Agar Strips; 25/Box
Representative image only.
EMD Millipore Mfr # 1442450025 – Item # EW-86509-62
Choose from strips with or without media
  • Prepared media strips make sampling effortless
PRODUCT OPTIONS
$195.00USD / Box of 25
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithBiotest Hycon RCS Air Sampler
  • DescriptionDG18 Agar Strips; 25/Box
Key Features
  • Prepared media strips make sampling effortless
More About this Item
Compatible with the Biotest Hycon RCS® Air Sampler 86509-00, the sampler operates on impaction principle, drawing airborne microorganisms onto an agar media strip for later incubation and analysis.
These agar strips come with Dichloran Glycerine Agar, ideal for determination of yeasts and molds.
Sampler sold separately.