EMD Millipore PLBC02510 Ultrafiltration Disc Membranes, 3k 25mm 10pk
Representative image only.
EMD Millipore Mfr # PLBC02510 – Item # EW-29949-10
Ideal for stirred cells and micropartition system
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
PRODUCT OPTIONS
$152.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)25
  • Nominal Molecular Weight Limit (Mwl) (Dalton)3000
  • Diameter (cm)2.5
  • DescriptionUltrafiltration Disc Membranes, 3k 25mm 10pk
Key Features
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
More About this Item
These high-recovery hydrophilic membranes are made of regenerated cellulose. Use when concentrating or desalting extremely dilute solutions or whenever your sample is hydrophobic.