Encon AQ120 Eyewash Refill Cartridge
Representative image only.
Encon Mfr # AQ120 – Item # EW-86001-41
Ensures safe, contaminant-free eye flushing
  • Self-evacuating cartridges maintain required flow for 15-minute flush without any external force
  • Once installed, side windows provide view of cartridge status
  • Sealed and ready-to-use
  • Complies with ANSI standards
$249.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionEyewash Refill Cartridge
Includes
Two individual bags of preserved solution.
Key Features
  • Self-evacuating cartridges maintain required flow for 15-minute flush without any external force
  • Once installed, side windows provide view of cartridge status
  • Sealed and ready-to-use
  • Complies with ANSI standards
More About this Item
For use with the Encon AQ100 emergency eyewash station, these sealed flexible cartridges have a 24 month shelf life that allows longer intervals of maintenance. There is no need to changed the solution out every few months like in standard self-contained eyewash stations. The cartridges provide contaminant free flushing that is safe, reliable, and pure. In one easy step, the eye wash station is pulled down and the self-evacuating cartridges begin to flow for the entire 15 minute flush.
Meets the ANSIZ358.1 Standard when installed in the Encon eyewash station (model 86001-40).
Not Available in Canada.