Endecotts ZSICSA-1.18 Sieve Calibration Standard, soda-lime glass, 16 mesh

Representative image only.
Endecotts Mfr # ZSICSA-1.18 – Item # EW-59945-04
Spheres are traceable to the United Kingdom National Physical Laboratory
  • Calibrate sieves to an accuracy of ±1µm
  • 6 month warranty

PRODUCT OPTIONS

$345.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • U.S. Standard Screen Size (No.)16
  • Tyler Standard Screen Size (mesh)14
  • Min Sieve Aperture (mm)1
  • Max Sieve Aperture (mm)1.4
  • Weight (g)50
  • DescriptionSieve Calibration Standard, soda-lime glass, 16 mesh
  • Warranty6 Months
6 Month WarrantyISO9001 Certified Supplier

Key Features

  • Calibrate sieves to an accuracy of ±1µm
  • 6 month warranty

More About this Item

Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture. Standards include a calibration chart.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat