Environmental Devices DS-10 Sampling Inlet; PM-10
Representative image only.
Environmental Devices Mfr # DS-10 PM-10 – Item # EW-10111-89
Optimize performance of your air/gas analyzer
  • Suitable for particulate matter size PM-10
PRODUCT OPTIONS
$243.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithDust-Sol dust sampler 10111-90
  • DescriptionSampling Inlet; PM-10
Key Features
  • Suitable for particulate matter size PM-10
More About this Item
Replacement inets, calibration equiptment, and other accessories to customize your sampling and analysis system.
This is compatible with Dust-Sol dust sampler 10111-90. Sampler sold separately.