Environmental Devices DS-2.5 Ds-2.5:Dust-Sol Pm-2.5 And Pm-10 Dust Sampler
Representative image only.
Environmental Devices Mfr # DS-2.5 – Item # EW-10111-90
Measure air quality impacts from a variety of sources, including volcanos, solid-fuel rocket engines and the oil fires
  • Lightweight and compact
  • Uses standard 47mm filter media
  • AC/DC power sources with low battery indicator
  • Programmable 7-day, 24 hour timer
  • 10,000 continuous hour diaphragm pump
  • Elapsed time indicator
  • Variable flow adjustment 0-6 LPM
  • 1 year warranty
$5,452.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDs-2.5:Dust-Sol Pm-2.5 And Pm-10 Dust Sampler
  • Warranty1 year
1 Year Warranty
Key Features
  • Lightweight and compact
  • Uses standard 47mm filter media
  • AC/DC power sources with low battery indicator
  • Programmable 7-day, 24 hour timer
  • 10,000 continuous hour diaphragm pump
  • Elapsed time indicator
  • Variable flow adjustment 0-6 LPM
  • 1 year warranty
More About this Item
The DS-2.5 PM-2.5 and PM-10 Dust Sampler is a device for monitoring PM-10 and PM-2.5 air. Small, durable, lightweight and battery operated, it is ideal for monitoring in remote areas or where no permanent sit has been established.
PM-10 & PM-2.5 EPA, Saturation Sampling: In saturation sampling, a network of samplers is deployed in an area to determine where the worst air quality problems exist. In addition to providing data for environmental impact studies, saturation sampling is frequently used to determine the location for permanent monitoring sites, define boundaries of air quality concerns and extent of point source impacts.