Environmental Devices EDC-7047 Filters For General Monitoring; 47 mm; Pk/25
Representative image only.
Environmental Devices Mfr # EDC-7047 – Item # EW-10112-03
Optimize performance of your air/gas analyzer
  • Ideal for general particulate monitoring
  • Tissuquartz 47-mm filters
PRODUCT OPTIONS
$90.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithDust-Sol dust sampler 10111-90
  • DescriptionFilters For General Monitoring; 47 mm; Pk/25
Conformite Europeenne
Key Features
  • Ideal for general particulate monitoring
  • Tissuquartz 47-mm filters
More About this Item
Replacement inets, calibration equiptment, and other accessories to customize your sampling and analysis system.
This is compatible with Dust-Sol dust sampler 10111-90. Sampler sold separately.