Environmental Devices EPAM-5000 Particulate Respiratory Health Safety Monitor
Representative image only.
Environmental Devices Mfr # EPAM-5000 – Item # EW-10111-96
Provides real-time data and gravimetric samples for analysis
  • Immediate display and data storage of lung-damaging aerosols— designed to investigate air for environmental, IAQ and industrial hygiene Reduces the number of manual gravimetric tests, as well as labor and analysis costs Unique aerodynamic particulate sizing and in-line filter cassette allows for concurrent gravimetric samplings Can complement all OSHA and EPA personal and ambient particulate air-sampling procedures Automatic clean air purging of sensor for increased stability and accuracy Internal temperature compensation for ambient use In-line gravimetric filter cassette Internal air sampling pump with adjustable flow rate Security code for protection of data integrity 4-key splashproof membrane and menu-driven operation Large data storage and compatible with Windows® NT, 2000/ME/XP— generate management-ready reports Housed in rugged, watertight carrying case Optional size-selective sampling inlets (1.0 ìm and 2.5 ìm cut points)
  • 1 year warranty
$8,096.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Particle Size (micron)0.1 to 100
  • Min Flow Rate (LPM)1
  • Max Flow Rate (LPM)4
  • DisplayLCD, 4 line, 20 character
  • Memory21,600 data points
  • OutputRS-232
  • PowerRechargeable battery
  • Battery Life24 hours
  • DescriptionParticulate Respiratory Health Safety Monitor
  • Warranty1 year
1 Year Warranty
Key Features
  • Immediate display and data storage of lung-damaging aerosols— designed to investigate air for environmental, IAQ and industrial hygiene Reduces the number of manual gravimetric tests, as well as labor and analysis costs Unique aerodynamic particulate sizing and in-line filter cassette allows for concurrent gravimetric samplings Can complement all OSHA and EPA personal and ambient particulate air-sampling procedures Automatic clean air purging of sensor for increased stability and accuracy Internal temperature compensation for ambient use In-line gravimetric filter cassette Internal air sampling pump with adjustable flow rate Security code for protection of data integrity 4-key splashproof membrane and menu-driven operation Large data storage and compatible with Windows® NT, 2000/ME/XP— generate management-ready reports Housed in rugged, watertight carrying case Optional size-selective sampling inlets (1.0 ìm and 2.5 ìm cut points)
  • 1 year warranty
More About this Item
Portable, microprocessor-based particulate monitor is suitable for ambient, environmental, and indoor air quality investigations. Highly sensitive, this unit scatters infrared light to measure particle concentration and provide immediate realtime determinations and data recordings of airborne particle concentration. The unique aerodynamic particle sizing and 47-mm in-line filter holder (optional accessory) loaded with the appropriate filter provide concurrent gravimetric sampling.