Environmental Express Alcohol Mix Std (9 Compts), 2000 µg/mL in DI Water; 1 mL

Representative image only.
Environmental Express Item # EW-97016-41
Multi-component organic standard for US EPA Method 8015
  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • 2,000 ppm alcohol mix organic standard in DI water matrix
  • 1 mL ampule

$52.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixDI Water
  • Element/IonAllyl alcohol, 1-Butanol, tert-Butyl alcohol, Ethanol, Ethylene glycol, Methanol, 2-Methyl-1-propanol, 1-Propanol, 2-Propanol
  • FormLiquid
  • Volume (mL)1
  • DOT InformationNot Regulated
  • Hazard SymbolHealth Hazard
  • UN NumberNot Regulated
  • CAS NumberAllyl alcohol (CAS #107-18-6), 1-Butanol (CAS #71-36-3), tert-Butyl alcohol (CAS #75-65-0), Ethanol (CAS #64-17-5), Ethylene glycol (CAS #107-21-1), Methanol (CAS #67-56-1), 2-Methyl-1-propanol (CAS #78-83-1), 1-Propanol (CAS #71-23-8), 2-Propanol (CAS #67-63-0)
  • Shelf Life (months)12
  • DescriptionAlcohol Mix Std (9 Compts), 2000 µg/mL in DI Water; 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • 2,000 ppm alcohol mix organic standard in DI water matrix
  • 1 mL ampule

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat