Environmental Express Barium, 1,000 µg/g for ICP in Mineral Oil; 100 g

Representative image only.
Environmental Express Item # EW-97066-19
Single-component metallo-organic standards for ICP analysis
  • ISO 9001:2015 certified and ISO/IEC 17025:2017 accredited
  • For ASTM Methods D7151, D5708, and D5185
  • 1,000 ppm barium metallo-organic standard in mineral oil matrix
  • 100 g bottle

PRODUCT OPTIONS

$120.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/g (1,000 ppm)
  • MatrixMineral Oil
  • Element/IonBarium (Ba)
  • FormLiquid
  • Weight (g)100
  • DOT InformationNot Regulated
  • Hazard SymbolHealth Hazard
  • UN NumberNot Regulated
  • Shelf Life (months)12

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified and ISO/IEC 17025:2017 accredited
  • For ASTM Methods D7151, D5708, and D5185
  • 1,000 ppm barium metallo-organic standard in mineral oil matrix
  • 100 g bottle

More About this Item

Use the metallo-organic standards for the determination of metals in oils and lubricants.

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat