Environmental Express BDXII Air Sampling Pump Starter Kit; 120 VAC
Representative image only.
Environmental Express Item # EW-60061-82
Convenient kits include everything you need for portable air sampling
  • Built-in rotameter and flow adjust offers flow rates of 500 to 3000 cc/min (0.50 to 3 LPM)
  • Compact, lightweight, and quiet sampling pump kits
  • Rechargeable battery pack system—ideal for portal use
  • Meets NIOSH 7400, 7082, 7105 and 7300
  • Single pump sampling starter kit
PRODUCT OPTIONS
$313.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Sampling pump1 BDXII Air Sampling Pump
  • DataloggingNo
  • ComplianceNIOSH 7400, 7082, 7105 and 7300
  • Battery TypeRechargeable NiMH Battery
  • Power (VAC)120
  • Power (Hz)60
  • DescriptionBDXII Air Sampling Pump Starter Kit; 120 VAC
Includes
one BDXII pump, battery, charger, tool kit, tubing, and clip assembly.
Key Features
  • Built-in rotameter and flow adjust offers flow rates of 500 to 3000 cc/min (0.50 to 3 LPM)
  • Compact, lightweight, and quiet sampling pump kits
  • Rechargeable battery pack system—ideal for portal use
  • Meets NIOSH 7400, 7082, 7105 and 7300
  • Single pump sampling starter kit
More About this Item
The BDXII Abatement Air Sampler is a dependable, low cost personal sampling pump. It is designed for use with asbestos and lead sampling. Meets NIOSH 7400, 7082, 7105 and 7300 specifications.
Choose between kits with one or five BDXII pumps with a Gilian or Zefon 5-station charger.