Environmental Express BPC1198 Packer Wide-Mouth Bottle, HDPE, Level 1, 250 mL, 1 mL Zinc acetate; 24/CS
Representative image only.
Environmental Express Item # EW-35207-76
Won't break like glass
  • Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory
  • Lightweight design with leakproof cap
  • Constructed of durable HDPE
PRODUCT OPTIONS
$14.75USD / Case of 500
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)250
  • MaterialHDPE
  • Cap / Stopper Size48/400
  • Cap ColorWhite
  • Bottle ColorNatural
  • Level Number1
  • Special Features1 mL Zinc acetate
  • Neck Inside Diameter (ID) (mm)48
  • DescriptionPacker Wide-Mouth Bottle, HDPE, Level 1, 250 mL, 1 mL Zinc acetate; 24/Cs
Key Features
  • Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory
  • Lightweight design with leakproof cap
  • Constructed of durable HDPE