Environmental Express Wide-Mouth Round Bottle, HDPE, Level 3, 1 gal; 4/CS
Representative image only.
Environmental Express Item # EW-35206-80
Won't break like glass
  • Provide easy-access wide-mouth
  • Lightweight design with leakproof cap
  • Included caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE
  • 3 month warranty
  • Model number BPC3150
PRODUCT OPTIONS
$23.80USD / Case of 4
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)3845
  • Capacity (L)3.845
  • Capacity (oz)128
  • Capacity (Gal)1
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialLDPE-lined Polypropylene
  • Cap / Stopper Size110 mm
  • Cap ColorWhite
  • Bottle ColorNatural
  • Special Features110 mm cap
  • AutoclavableNo
  • Level Number3
  • DescriptionPre-Cleaned Wide-Mouth Round Bottle, HDPE, Level 3, 1 gal; 4/Cs
  • Warranty3 month
3 Month Warranty
Key Features
  • Provide easy-access wide-mouth
  • Lightweight design with leakproof cap
  • Included caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE
  • 3 month warranty
  • Model number BPC3150
More About this Item
Choose between Level 1 or 3 pre-cleaned round wide-mouth bottles. Level 1 bottles are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory. Level 3 bottles meet EPA guidelines, but do not ship with a certificate of analysis. They are ideal for general-use.