Environmental Express C7125 SimpleDist® Snip & Pour Reagent, 20 g/L Boric Acid, 5 mL Tube; 50/PK

Representative image only.
Environmental Express Item # UX-35209-83
Reagent addition is a snap with Snip & Pour reagent tubes
  • Eliminate measuring and pipetting
  • Increase accuracy
  • Improve quality control and final results
  • Meet DOT requirements
  • 3 month warranty

PRODUCT OPTIONS

$92.10USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaH3BO3
  • ClassTrapping Solution
  • SynonymsBoric acid
  • FormLiquid
  • Volume (mL)250
  • Weight (mL)250
  • Packaging250 mL Total, 5 mL individual PTFE ampules
  • CAS Number10043-35-3
  • DescriptionSimpleDist® Snip & Pour Reagent, 20 g/L Boric Acid, 5 mL Tube; 50/PK
  • Warranty3 month
3 Month Warranty

Key Features

  • Eliminate measuring and pipetting
  • Increase accuracy
  • Improve quality control and final results
  • Meet DOT requirements
  • 3 month warranty

More About this Item

Snip & Pour concentrates are packaged in convenient PTFE tubes. Just snip the corner of the PTFE tube and pour the contents directly into SimpleDist® collection trap. Premeasured reagents are meant for use with cyanide or ammonia distillation.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat