Environmental Express Cole Parmer P200-01 pH Meter Kit; 3-in-1 pH Electrode

Representative image only.
Environmental Express Item # EW-59250-00
Options for every application
  • Choice of electrodes—combination 3-in-1 with ATC or Tris-compatible

PRODUCT OPTIONS

$502.00USD / Each

$164.50 - $237.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Accessories Today!

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01, 0.1
  • pH Accuracy±0.01
  • Buffer RecognitionUSA, NIST, user defined
  • Calibration1 to 3 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Accuracy±0.01% full-scale
  • Max Conductivity
  • Min Temperature (° C)0
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1°F/C
  • Temperature Accuracy±0.36°F (0.2°C)
  • Temperature compensationYes, 32 to 212°F (0 to 100°C), automatic or manual
  • Power (VAC)110 - 120
  • Power (Hz)50/60
  • DescriptionP200-01 pH Meter Kit; 3-in-1 pH Electrode
3 Year Warranty

Includes

pH electrode and power cord.

Key Features

  • Choice of electrodes—combination 3-in-1 with ATC or Tris-compatible

More About this Item

The P200 series of benchtop meters provides accurate, reliable results with the benefit of convenient accessories to make your lab run smoothly. Large color TFT display is easily visible, and features a stability icon to alert you when your readings are ready to be recorded and an electrode health icon indicating when you need to refill or replace your electrode.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat