Environmental Express Color-Coded Surrogate, BN/A (100:100), for CLP; 25 mL

Representative image only.
Environmental Express Item # EW-35200-69
Prevent costly mistakes by color coding your extraction fluids
  • Ideal for semi-volatile extractions like EPA Methods 3500C, 625, and 8270
  • Simply add 1 mL of the methanol based indicator to your sample
  • Store in your lab refrigerator

PRODUCT OPTIONS

$67.80USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ClassGeneral Solutions
  • FormLiquid
  • Volume (mL)25
  • Packaging25 mL
  • DOT InformationNot regulated by DOT
  • Storage SymbolRefrigerate
  • Hazard SymbolFlammable, Toxic, Organ damage
  • CAS Number67-56-1, 4165-60-0, 367-12-4, 4165-62-2, 321-60-8, 1718-51-0, 118-79-6, 2199-69-1, and 93951-73-6
  • Shelf Life (months)30 (refrigeration required)
  • DescriptionColor-Coded Surrogate, Base Neutral/Acid (100:100), for CLP; 25 mL

Key Features

  • Ideal for semi-volatile extractions like EPA Methods 3500C, 625, and 8270
  • Simply add 1 mL of the methanol based indicator to your sample
  • Store in your lab refrigerator

More About this Item

Color coded surrogates are methanol based and supplied in 25mL volumes in screw-top vials — add 1 mL to your sample and help avoid performing re-extractions. These surrogates act as an indicator to ensure the pH range of the sample is correct.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat