Environmental Express E083700C Cubitainer, LDPE, 20 Liter; 36/Cs
Representative image only.
Environmental Express Item # EW-35204-86
Collapsible containers nest for efficient storage
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)20000
  • Capacity (L)20
  • Capacity (oz)676
  • Capacity (Gal)5
  • MaterialLow Density Polyethyene (LDPE)
  • Bottle ColorNatural
  • AutoclavableNo
  • DescriptionCubitainer, LDPE, 20 Liter; 36/Cs
  • Warranty3 month
3 Month Warranty
Key Features
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
  • 3 month warranty
More About this Item
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.
Natural LDPE containers come assembled with LDPE-lined size 38 caps.