Environmental Express Pre-Cleaned Round Narrow-Mouth Bottle, HDPE, Level 1, 500 mL; 24/CS

Representative image only.
Environmental Express Item # EW-35206-32
Stand up to your demands
  • Easy-pour design allows for simple handling
  • 3 month warranty
  • Model number BPC1010

PRODUCT OPTIONS

$17.30USD / Case of 24

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)500
  • Capacity (L)0.5
  • Capacity (oz)16
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialLDPE-lined Polypropylene
  • Cap / Stopper Size24 mm
  • Cap ColorWhite
  • Bottle ColorNatural
  • AutoclavableNo
  • Level Number1
  • DescriptionPre-Cleaned Round Narrow-Mouth Bottle, HDPE, Level 1, 500 mL; 24/Cs
  • Warranty3 month
3 Month Warranty

Key Features

  • Easy-pour design allows for simple handling
  • 3 month warranty
  • Model number BPC1010

More About this Item

These pre-cleaned round narrow-mouth bottles come in two levels. Level 1 bottles are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory.
Level 3 bottles meet EPA guidelines, but do not ship with a certificate of analysis—ideal for general-use.

Included caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat