Environmental Express D1053 BOD Bottle Storage Rack, Epoxy-Coated Steel, 300 mL, 12-place
Representative image only.
Environmental Express Item # EW-53200-23
Safe and convenient organization of BOD bottles
  • Stackable
  • Choice of polypropylene or steel construction
  • 3 month warranty
PRODUCT OPTIONS
$135.75USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialEpoxy-coated steel
  • Length (in)13
  • DescriptionBOD Bottle Storage Rack, Epoxy-Coated Steel, 300 mL, 12-place
  • Warranty3 month
3 Month Warranty
Includes
Racks do not include BOD bottles.
Key Features
  • Stackable
  • Choice of polypropylene or steel construction
  • 3 month warranty
More About this Item
Polypropylene racks can be autoclaved up to 121°C. Steel racks are more durable but cannot be autoclaved.
These stackable, rugged wire racks are plastic-coated to avoid corrosion.