Environmental Express G1090 StepSaver Extraction Station, 90 mm, Glass Funnel; 1/Ea
Representative image only.
Environmental Express Item # EW-35210-07
Individual glass stations for oil and grease analysis
  • Option of 47- or 90-mm glass funnels; or no funnel
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • DescriptionStepSaver Extraction Station, 90 mm, Glass Funnel; 1/Ea
  • Warranty3 month
3 Month Warranty
Key Features
  • Option of 47- or 90-mm glass funnels; or no funnel
  • 3 month warranty
More About this Item
The improved StepSaver™ is completely manufactured out of durable PVC and plastic materials. This compact, single-place station has a small footprint to save valuable benchtop space. These stations can be daisy-chained together to run multiple samples at once for a faster sample throughput.  The improved StepSaver still meets the EPA Method 1664 requirements for Oil and Grease extractions. 