Environmental Express GhostWipe® SC4256 Lead Wipe Standard, 175-250 ug in 100 mL Cup; 10/Pk
Representative image only.
Environmental Express Item # EW-35208-79
Simplify your surface sample collection and preparation for lead and beryllium content
  • Just wipe surface you want to test, digest, and analyze results
  • Wipes breakdown completely and wont clog equipment
  • Complies with ASTM method E 1792 for lead or beryllium in dust
  • Individually packaged in a 100 mL digestion cup — ready for reagents and to be processed in a HotBlock System
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • TestLead standard
  • Tests Per Kit10
  • DescriptionLead Wipe Standard, 175-250 ug in 100 mL Cup; 10/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Just wipe surface you want to test, digest, and analyze results
  • Wipes breakdown completely and wont clog equipment
  • Complies with ASTM method E 1792 for lead or beryllium in dust
  • Individually packaged in a 100 mL digestion cup — ready for reagents and to be processed in a HotBlock System
  • 3 month warranty
More About this Item
The Ghostwipe completely dissolves during the digestion procedure thus allowing
everything into solution. Dependent upon the material wiped, filtration may not
be necessary. In addition, the Ghostwipe is more rugged than other traditional
paper wipes and will not fall apart during the wiping of rough surfaces.
Each Spike sample contains 175-250 ug of Lead per wipe. Using a NIST-Traceable solid sample, known amounts are placed onto a GhostWipe and individually labeled with the exact amount of lead each wipe contains.