Environmental Express HotBlock® 200 Programmable Temperature Controller, Single Block; 1/Ea

Representative image only.
Environmental Express Item # UX-53206-00
Control one or two block heaters
  • Choose your controller, then block (or blocks) to suit your metals digestion needs
  • Mount controllers outside the fume hood on metal surfaces, in an available outlet, or place on a benchtop
  • Same dependability with increased temperature range
  • 1 year warranty

PRODUCT OPTIONS

$1,284.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Number Of Blocks1
  • Uniformity (° C)±2.0°
  • Max Temperature (° C)200
  • Power (VAC)120
  • Width (in)6
  • Length (in)7
  • DescriptionProgrammable Temperature Controller, Single Block; 1/Ea
  • Warranty1 year
1 Year Warranty

Key Features

  • Choose your controller, then block (or blocks) to suit your metals digestion needs
  • Mount controllers outside the fume hood on metal surfaces, in an available outlet, or place on a benchtop
  • Same dependability with increased temperature range
  • 1 year warranty

More About this Item

Blocks and controllers are sold separately.
Perform a wider range of experiments and digestions with the expanded temperature range—up to 200°C* with the new HotBlock 200! Controllers feature automatic shut-off, alarms, and ability to save profiles with ramp and soak settings. The dual-block controller can control up to two blocks independently and simultaneously. Safely place the external controller outside the fume hood. Controllers may be mounted to a metal surface using the magnets, or on an electrical outlet.

*Use glass or PTFE digestion cups when operating above 150°C.

PART OF A COMPLETE SYSTEM – GET EVERYTHING YOU NEED

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat