This item is now discontinued. Please browse similar items
Environmental Express HotBlock Soil-Cell Extraction Cell Inner Lid, Stainless Steel; Each
Representative image only.
Environmental Express Item # EW-35208-84
Save time and money with the Soil-Cell extraction system
  • Reduce amount of solvent that is used compared to traditional methods
  • Quick 30 minute extraction saves time
  • Closed-cell technology prevents contamination
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Compatible WithSoil-Cell System
  • DescriptionEnvironmental Express HotBlock Soil-Cell Extraction Cell Inner Lid, Stainless Steel; Each
Key Features
  • Reduce amount of solvent that is used compared to traditional methods
  • Quick 30 minute extraction saves time
  • Closed-cell technology prevents contamination
More About this Item
The Soil-Cell system is developed for the extraction of semi-volatile organic compounds in soil samples. The Soil-Cell system meets the requirements of the Performance Based Measurement System established by the EPA and replaces microwave technology with a HotBlock and stainless steel Soil Extraction Cells. Analytical results of a CRM (Certified Reference Material) for PAH/BNA and TPH fell within the required acceptance limits and were very reproducible. Real world soil samples were also extracted using this procedure for PAH/BNA, TPH, and PCB compounds and compared to results achieved using SW846 Method 3545A. Results were comparable for all three groups of compounds.
Replacement extraction cell stainless steel inner lid (cell body, O-rings, and threaded cap sold separately).