Environmental Express MCE Air Analysis Filters, 1 µg/filter of BeO as Be

Representative image only.
Environmental Express Item # EW-97016-50
Ensure worker safety by monitoring workplace air quality
  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3105
  • Designed to meet QC requirements for NIOSH Method 7300 and US EPA Method SW-846
  • 37 mm diameter
  • 0.8 µm pore size

PRODUCT OPTIONS

$106.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1 µg/filter
  • MatrixMixed Cellulose Ester
  • Element/IonBeryllium Oxide as Beryllium
  • DOT InformationNot Regulated
  • Hazard SymbolNot Regulated
  • UN NumberNot Regulated
  • Shelf Life (months)12
  • DescriptionMCE Air Analysis Filters, 1 µg/filter of BeO as Be

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3105
  • Designed to meet QC requirements for NIOSH Method 7300 and US EPA Method SW-846
  • 37 mm diameter
  • 0.8 µm pore size
  • 1 µg/filter of beryllium oxide particulates on MCE filters
  • Two loaded filters

More About this Item

Beryllium Oxide is a toxic particulate material present in workplace samples that can be harmful to workers. Use these filters for method development or quality control checks to ensure worker safety. The source of the Beryllium Oxide applied to the filter is NIST SRM 1877 high-fired BeO powder. Blanks are supplied separately; contact us for information

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat