Environmental Express Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 35 mg Sodium Thiosulfate Preserved, 125 mL; 48/CS
Representative image only.
Environmental Express Item # EW-35209-44
Level 1 bottles are certified and come with a certificate of analysis
  • Preserved and unpreserved bottles available
PRODUCT OPTIONS
$84.80USD / Case of 48
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)125
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • Special Features35 mg sodium thiosulfate preserved
  • Level Number1
  • Neck Inside Diameter (ID) (mm)38
  • DescriptionLevel 1 Certified Natural Color Bottle, Round, Wide Mouth, 35 mg Sodium Thiosulfate Preserved, 125 mL, 48/Case
Key Features
  • Preserved and unpreserved bottles available
More About this Item
These HDPE bottles are Level 1 certified to meet the EPA specifications and guidance for contaminant free sample collection containers.