Environmental Express QC-TMFM-X Trace Metals Filters, 1 to 10 µg, 37 mm; 10 filters, 5 blanks SKU 3520900
Representative image only.
Environmental Express Item # EW-35209-00
Added quality control to your industrial hygiene testing
  • Meets NIOSH Method 7300 requirements
  • Industrial hygiene standards for trace metals
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTrace Metals Filters, 1 to 10 µg, 37 mm; 10 filters, 5 blanks
  • Warranty3 month
3 Month Warranty
Key Features
  • Meets NIOSH Method 7300 requirements
  • Industrial hygiene standards for trace metals
  • 3 month warranty
More About this Item
Filters are 37-mm diameter mixed cellulose ester (MCE) with a 0.8 µm pore size. The concentrations are reported as µg per filter, and are certified to ±1%. Expiration date of one year.
Trace metal in µg per filter
Aluminum -
Arsenic 10
Barium 2.5
Beryllium 1.0
Cadmium 1.0
Chromium 2.5
Cobalt 2.5
Copper 2.5
Iron 2.5
Lead 2.5
Maganese 1.0
Nickel 2.5
Silver 1.0
Thalium 2.5
Vanadium 2.5
Zinc 2.5
Uranium -
Available for sale in the United States, UK, and India only