Environmental Express SimpleWater BOD Nutrient Buffer Solution, 19L; 36/Pk

Representative image only.
Environmental Express Item # EW-53200-09
No mess--simply pour contents into your container and add water
  • Range of five standard concentrations to make a volume of 300 mL to 19 L
  • Borosilicate glass vials with leakproof screw caps

PRODUCT OPTIONS

$61.70USD / Pkg of 36

SPECS & DESCRIPTION

Specifications & Description

  • ClassBOD Dilution Water Concentrate
  • SynonymsBOD dilution water
  • FormLiquid
  • Volume (mL)19000
  • Weight (mL)19000
  • Packaging19000 mL For 19 L dilution
  • CAS Number12125-02-9, 10035-04-8, 10025-77-1, 10034-99-8, 7758-11-4, 7778-77-0, and 7558-79-4
  • DescriptionBOD Nutrient Buffer Solution, 19L; 36/Pk

Key Features

  • Range of five standard concentrations to make a volume of 300 mL to 19 L
  • Borosilicate glass vials with leakproof screw caps

More About this Item

BOD dilution water is composed of a variety of nutrients that provide optimal growth media for bacteria in BOD tests. SimpleWater nutrient buffer solutions are concentrates of the four nutrient solutions conveniently combined into one vial. Simply dilute the contents of vial to the specified volume, and you've got your BOD dilution water!

Available in five standard concentrations, borosilicate glass vials with leakproof screwcaps. Vials in pack of 36.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat