Environmental Express Snip & Pour® Glucose-Glutamic Acid (GGA), 24 x 6 mL Vials/PK
Representative image only.
Environmental Express Item # EW-53200-11
Eliminate pipetting and dilutions
  • No more worries about sample contamination
  • Ready-to-use dilutions are easy and convenient
  • No pipetting, no dilution factor, and no contamination = lower operating cost
  • 3 month warranty
PRODUCT OPTIONS
$83.50USD / Pkg of 24
SPECS & DESCRIPTION
Specifications & Description
  • Buffer Kit Values24 vials
  • Volume (mL)6
  • DescriptionSnip & Pour® Glucose-Glutamic Acid (GGA), 24 x 6 mL Vials/PK
  • Warranty3 month
3 Month Warranty
Key Features
  • No more worries about sample contamination
  • Ready-to-use dilutions are easy and convenient
  • No pipetting, no dilution factor, and no contamination = lower operating cost
  • 3 month warranty
More About this Item
Each vial contains the APHA specified concentration of GGA. Simply shake the vial thoroughly, remove the seal and cap, pour contents, and rinse the vial at least two times with BOD water into the check standard solution. Product has a 24-month shelf life.
GGA Ampulesstable, certified single use ampules for easy GGA test preparations for 300 mL bottles. Twenty-four 6 mL vials per package.