Environmental Express Snip & Pour Lab Control Standards, 40mg HEM/10mL Ace.; 20/Pk

Representative image only.
Environmental Express Item # EW-35200-22
Premeasured controls for oil and grease analysis
  • Meets requirements for EPA Method 1664
  • Each lot is quality checked gravitmetrically and by GC
  • No pipetting or warming needed—just snip a corner and pour

PRODUCT OPTIONS

$111.32USD / Pkg of 20

SPECS & DESCRIPTION

Specifications & Description

  • ClassGeneral Solutions
  • ChemistryAcids;Analytical Reagents;Inorganics;Solutions;Solvents;Standards
  • FormLiquid
  • Volume (mL)200
  • Weight (mL)200
  • Packaging200 mL total (pack of 20 x 10 mL vials)
  • DOT InformationAcetone; Class 3, Packing Group II
  • CAS Number67-64-1, 544-76-3, and 57-11-4
  • UN NumberUN1090
  • Shelf Life (months)36
  • Description40 mg HEM in 10 mL Acetone; 20/Pk

Includes

Certificate of Analysis and SDS.

Key Features

  • Meets requirements for EPA Method 1664
  • Each lot is quality checked gravitmetrically and by GC
  • No pipetting or warming needed—just snip a corner and pour

More About this Item

Convenient Snip & Pour standards meet all requirements for EPA Methods 1664 (paragraph 7.10) Hexadecane/Stearic Acid Spiking Solution for oil and grease extraction, and total petroleum hydrocarbons testing. The single-use tubes deliver a precise volume of hexane extractable material (HEM) to your Laboratory Control (LCS) or Matrix Spike samples.
The PTFE tubes allow you to easily dispense without a pipette and won't retain fluid—simply cut a corner and pour the contents. All laboratory controls include a Certificate of Analysis with lot number, gravimetric concentration, and expiration date, as well as an SDS.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat