Environmental Express SPE-Express® 2 47 mm Disk, Individual Station, 120/240 VAC
Representative image only.
Environmental Express Item # EW-35390-14
Choose from a variety of additional accessories to expand and customize your system
  • Can string up to 3 stations together and operate simultaneously
  • 1 year warranty parts & labor
PRODUCT OPTIONS
$17,000.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Description47 mm Disk, Individual Station, 120/240 VAC
  • Warranty1 year parts & labor
1 Year Parts and Labor
Key Features
  • Can string up to 3 stations together and operate simultaneously
  • 1 year warranty parts & labor
More About this Item
The SPE-Express® 2 system is intended for the automated extraction of oil and grease material following US EPA Method 1664. The self-contained unit employs a Solid Phase Extraction (SPE) disk to capture and separate oil and grease components from sample media. Components are eluted from the SPE disk using n-hexane and collected in an aluminum pan. Following elution and collection, the remaining solvent mixture is then evaporated through gently applied heat source removing the n-hexane and capturing the oil and grease component residue.