Environmental Express StepSaver System, 7-Station, PVC/Glass; Ea
Representative image only.
Environmental Express Item # EW-35210-02
Solid phase extraction systems simplify EPA Method 1664 for oil and grease
  • Ready to be evaporated without a separate drying step
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Filter Diameter (mm)47
  • Body MaterialGlass, PVC
  • DescriptionSystem, 7-Station, PVC/Glass; Ea
  • Warranty1 year
1 Year Warranty
Key Features
  • Ready to be evaporated without a separate drying step
  • 1 year warranty
More About this Item
StepSaver systems streamline the extraction process by allowing the analyst to collect the solvent with eluted oil and grease directly into a collection flask. As fluid enters the collection flask, it passes through a drying cartridge to remove excess water. It is ready to be evaporated without a separate drying step.
7-Station PVC Manifold
Each station comes with:
1 1000mL glass funnel
1 extraction head with 40/35 ground glass joint
1 120° 2-way PTFE stopcock
1 stainless steel filter screen
1 flourosilicone sealing gasket
1 aluminum clamp
1 Keck clip
2 100mL borosilicate glass collection flasks
1 4-Liter HDPE waste carboy with tubing and fittings
Consumables:
2 boxes of 20 47 mm UltraFlow Filters
1 pack of 50 sodium sulfate drying cartridges
1 box of 20 x 10mL 40mg HEM Snip and Pour standards
1 box of 20 x 10mL 7 mg HEM MDL Snip and Pour standards
Available for sale in the United States, UK, and India only