Environmental Express Straight-Side Glass Jar, Level 1, Amber, 60 mL; 24/CS

Representative image only.
Environmental Express Item # UX-35204-00
Get to your samples easier
  • Available with PTFE or PE foam-lined caps
  • Select amber glass bottles for light-sensitive samples
  • 3 month warranty
  • Model number APC1265

PRODUCT OPTIONS

$41.00USD / Case of 24
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)60
  • Capacity (L)0.06
  • Capacity (oz)2
  • MaterialGlass
  • Cap MaterialPTFE-lined Polypropylene
  • Cap / Stopper Size53 mm
  • Bottle ColorAmber
  • AutoclavableNo
  • Special FeaturesPTFE-lined Polypropylene Cap
  • Level Number1
  • DescriptionStraight-Side Glass Jar, Level 1, Amber, 60 mL; 24/Cs
  • Warranty3 month
3 Month Warranty

Key Features

  • Available with PTFE or PE foam-lined caps
  • Select amber glass bottles for light-sensitive samples
  • 3 month warranty
  • Model number APC1265

More About this Item

Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory. Included caps are composed of polypropylene (PP) and lined with PTFE.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat