Environmental Express T80WL Portable White Light Turbidity Meter Kit
Representative image only.
Environmental Express Item # UX-35635-21
Easily measure water sample turbidity in the field or on the bench
  • Rugged, portable design adapts to use in the field or on your lab bench
  • Simplistic user interface for easy operation at any skill level
  • Standards included as a kit so your technicians are ready to calibrate and measure right out of the box!
  • Large TFT color screen for easy operation
  • Improved accuracy with average measurement mode
  • Battery operated and IP67-rated housing—ideal for use in the field
  • Seamless method compliance for U.S. EPA method 180.1
  • Also available in an Infrared configuration (part number: 35635-22 for compliance with European standard ISO 7027 (DIN EN 27027) method (source water analysis)
PRODUCT OPTIONS
$1,085.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeTurbidity
  • Display typeTFT color screen
  • ConnectionsUSB
  • CalibrationYes
  • Min Turbidity0 NTU
  • Max Turbidity1000 NTU
  • Turbidity Resolution0.01/0.1/1 NTU (FNU)
  • Turbidity Accuracy≤ ±2 % of reading + stray light
  • WaterproofIP67
  • Battery1 rechargeable lithium-ion (included)
  • Battery life (hr)20
  • Accessories IncludedCalibration solutions (0.0, 20.0, 100, 800 NTU), carrying case, microfiber cloth, power adapter (5V 1A), silicone oil (10 mL), USB cable, six sample vials, and user manual.
  • DescriptionPortable White Light Turbidity Meter Kit
  • Warranty2 years
Two year warrantyConformite Europeenne
Includes
Turbidimeter, calibration solutions (0.0, 20.0, 100, 800 NTU), carrying case, microfiber cloth, 1 rechargeable lithium-ion battery, power adapter (5V 1A), silicone oil (10 mL), USB cable, six sample vials, and user manual.
Key Features
  • Rugged, portable design adapts to use in the field or on your lab bench
  • Simplistic user interface for easy operation at any skill level
  • Standards included as a kit so your technicians are ready to calibrate and measure right out of the box!
  • Large TFT color screen for easy operation
  • Improved accuracy with average measurement mode
  • Battery operated and IP67-rated housing—ideal for use in the field
  • Seamless method compliance for U.S. EPA method 180.1
  • Also available in an Infrared configuration (part number: 35635-22 for compliance with European standard ISO 7027 (DIN EN 27027) method (source water analysis)
More About this Item
US EPA Safe drinking water regulations require measurement of turbidity every 15 minutes. Wastewater facilities require turbidity measurement before and after filter bed backflush. In any lab or field setting, Environmental Express turbidity meters provide adaptability needed to easily complete timely, error free testing requirements.
Our portable turbidity meters have a rugged waterproof, IP67-rated design suitable for use in all environments. Automatic functions like “continuous” measurement mode generate maximum, and minimum measured values for multiple readings as well as averaging for ≥10 readings. Automatic accurate measurement and data treatments means testing schedules are easily maintained without disruption.