Eppendorf 22-60-000-1 Microcentrifuge Tube, 0.5 mL, Natural Color
Representative image only.
Eppendorf Mfr # 22-60-000-1 – Item # EW-02560-32
$36.00USD / Pkg of 50
BUY MORE & SAVE
  • 1-5$36.00USD / Pkg of 50
  • 6-29$32.40USD / Pkg of 50
  • 30-119$30.60USD / Pkg of 50
  • 120+$28.80USD / Pkg of 50
Often Bought with…
Ohaus Accessory Carrying Case for CS/CL Scales
$53.69USD / Each
$59.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g25000
  • SterileYes
  • AutoclavableYes
  • Tube Capacity (mL)0.5
  • Tube MaterialPolypropylene
  • Tube ColorNatural
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleClick-Fit
  • Cap MaterialPolypropylene
  • GraduationsNone
  • DescriptionMicrocentrifuge Tube, 0.5 mL, Natural Color
More About this Item
Eppendorf® Biopur™ tubes have all the features of the standard and Safe-Lock™ tubes and are individually wrapped and certified sterile. Biopur™ tubes are pyrogen-, RNase-, DNA-and ATP-free; supplied with a quality assurance card.