Eppendorf 5425 Ventilated Microcentrifuge with 24-Tube Rotor; 120V, 50/60 Hz

Representative image only.
Eppendorf Mfr # 5405000042 – Item # EW-02580-00
Added versatility to spin microcentrifuge tubes, miniprep spin columns, and PCR tube strips up to 15,000 rpm
  • Lid automatically opens at end of run for easy sample removal
  • Suitable for use in cold rooms
  • Whisper-quiet operation

$3,386.00USD / Each

$161.00USD / Each

for an additional
$161.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Max Speed (rpm)15000
  • Max RCF x g21130
  • Speed ControlTouch Pad
  • Rotor TypeFixed-Angle
  • Rotors IncludedYes
  • Noise level (dB(A))<51
  • Max Capacity (mL)48
  • Tube Capacity24 x 1.5/2.0 mL
  • Length (in)8
  • Width (in)9 13/64
  • Height (in)12
  • Length (cm)22.5
  • Width (cm)23.4
  • Height (cm)32
  • Height With Lid Open (in)17
  • Height With Lid Open (cm)44
  • Power (VAC)120
  • Power (Hz)50/60
  • DescriptionVentilated Microcentrifuge with 24-tube rotor - 120V, 50/60 Hz
Two year warranty

Includes

24 x 1.5/2.0-mL fixed-angle rotor with aeresol-tight lid and power cord with plug.

Key Features

  • Lid automatically opens at end of run for easy sample removal
  • Suitable for use in cold rooms
  • Whisper-quiet operation

More About this Item

Unit features microprocessor-based control with five-digit speed display of time and speed/g force. Easy-to-access control options include short-spin speed setting, soft-braking, “At Set” timer function, and adjustable volume alarm. Smooth keypad provides easy wipe-down cleaning. Aerosol-tight rotor is suitable even for hazardous materials, and contoured bowl design minimizes noise. Order optional rotors, below right, for added versatility.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat