Eppendorf 940001102 Cuvette stand for 16 Uvettes
Representative image only.
Eppendorf Mfr # 940001102 – Item # EW-83600-55
Designed for easy and efficient molecular analyses
  • Optimize performance of your Eppendorf spectrophotometer with these accessories
  • ¡Integrated Peltier element for temperature control of the cuvette shaft
  • Freely selectable cuvette shaft temperature control in a range of +20°C to +42°C with 0.1°C increments
  • Preprogrammed and freely programmable kinetic methods
  • Subsequent modification of the time window for regression evaluation
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCuvette stand for 16 Uvettes
Key Features
  • Optimize performance of your Eppendorf spectrophotometer with these accessories
  • ¡Integrated Peltier element for temperature control of the cuvette shaft
  • Freely selectable cuvette shaft temperature control in a range of +20°C to +42°C with 0.1°C increments
  • Preprogrammed and freely programmable kinetic methods
  • Subsequent modification of the time window for regression evaluation
More About this Item
Improve your molecular anayses with these quality accessories.