Eppendorf epTIPS Single Use Racks 1250 uL 480/Pk
Representative image only.
Eppendorf Mfr # 0030075269 – Item # EW-18888-14
High standards of purity ideally suited for critical laboratory applications
  • Pyrogen-free (endotoxin-free) according to LAL test
  • Sterilized by beta radiation to SAL 10-6, avoiding contamination by ethylene oxide
  • Tested and certified by an independent laboratory
  • Choose Biopur tips for molecular and microbiology, cell technology, quality control, and forensic applications
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)1250
  • Min Volume (µL)50
  • Max Volume (mL)1.25
  • Tip Volume (mL)1.25
  • Max Dispensing (mL)1.25
  • Package OfIndividually Wrapped Single Use Racks; 480/Pk
  • DescriptionepTIPS Single Use Racks 1250 uL 480/Pk
Key Features
  • Pyrogen-free (endotoxin-free) according to LAL test
  • Sterilized by beta radiation to SAL 10-6, avoiding contamination by ethylene oxide
  • Tested and certified by an independent laboratory
  • Choose Biopur tips for molecular and microbiology, cell technology, quality control, and forensic applications
More About this Item
Pipette tips sterilized by beta radiation maintain purity greater than those exposed to ethylene oxide, which is a gas absorbed by plastics.
Additional certifications for Biopur tips include DNA-free, DNase-free, RNase-free, PCR inhibitor-free, and ATP-free.