Erie Scientific 2951-4-001 Clipped-Corner Slides, frosted glass, 144/PK
Representative image only.
Erie Scientific Mfr # 2951-4-001 – Item # EW-48520-03
Corners are clipped for easy handing
PRODUCT OPTIONS
$67.50USD / Pkg of 144
SPECS & DESCRIPTION
Specifications & Description
  • Slide TypeFrosted
  • DescriptionClipped-Corner Slides, frosted glass, 144/PK
More About this Item
Drastically reduce the chances of cutting your finger or tearing a glove when you use these slides. Clipped corners keep sample away from the edges of the slide—ideal for making smears. Slides measure 75 x 25 mm and are 1-mm thick. Available in one gross (144 slides).