Erie Scientific Colorfrost® Plus Adhesion Microscope Slides, Pink; 144/Pk

Representative image only.
Erie Scientific Mfr # 7951PLUS-001 – Item # EW-48512-08
Tissue sections and cells electrostatically adhere to the glass without adhesives – eliminates tissue loss during staining
  • Color-coded for quick and simple identification
  • Marking surfaces ensure easy labeling
  • Slides measure 75 x 25 mm and are 1-mm thick
  • 1 year warranty
Often Bought With…

SPECS & DESCRIPTION

Specifications & Description

  • Slide TypePink Colorfrost Plus
  • DescriptionPink Colorfrost Plus Adhesion Slides, 144/pk
  • Warranty1 year
1 Year Warranty

Key Features

  • Color-coded for quick and simple identification
  • Marking surfaces ensure easy labeling
  • Slides measure 75 x 25 mm and are 1-mm thick
  • 1 year warranty

More About this Item

A permanent positive charge applied to the slide surface allows slides to hold tissue sections without the need for expensive or time-consuming preparatory steps in the laboratory

This treatment virtually eliminates tissue loss during staining and electrostatically attracts frozen tissue sections and cytology preparations, binding them to the slide.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat