ETCON WL140 Wire Length Meter with Leads
Representative image only.
ETCON Mfr # WL140 – Item # EW-20633-45
$1,511.50USD / Each
Often Bought with...
Cole-Parmer ThinTouch™ Nitrile Gloves
$9.00 - $67.50USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWire Length Meter with Leads
More About this Item
Measures any reel of wire from 24 AWG to 1000MCM stranded and solid in lengths of 15ft to 20,000ft. The unit is useful for electrical contractors, utilities and distributors for determining inventories of wire. The operator selects gauge, aught or MCM mode, selects copper or aluminum mode, attaches the alligator clips to the ends of the wire being measured, and presses {length} button to determine the length of wire.
FEATURES
  • Microprocessor based meter designed to an accuracy of ±1% with a 4-1/2 digit LCD readout
  • Direct readout for both copper and aluminum wire
  • Automatic internal adjustment for ambient temperature
  • Unique calibration mode enables measurement of non-standard gauge wire
  • Only one alligator clip attachment required at each end of wire
  • Field replacement of batteries
  • Insulation piercing probe assemblies are included enabling use without stripping insulation
  • Impact resistant carrying case, alligator clip assemblies, insulation piercing assemblies, batteries and new probe leads included
  • Made in the U.S.A.