Eurotherm V560 Current Indicator, Loop Powered, 4-20 or 10-50 mA DC
Representative image only.
Eurotherm-Action Instruments Mfr # V560-0014-0 – Item # EW-68478-51
  • Eliminates extra power supply
  • LCD readable from 30 ft
  • Switch selectable mA inputs and decimal points
  • Readings to ±19990
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Display3.5\" Digit LCD
  • Output4-20 mA
  • DescriptionCurrent Indicator, loop powered, 4-20 or 10-50 mA DC
  • Warranty3 year
3 Year Warranty
Key Features
  • Eliminates extra power supply
  • LCD readable from 30 ft
  • Switch selectable mA inputs and decimal points
  • Readings to ±19990
  • 3 year warranty
More About this Item
Intelligent, universal process two-wire transmitters feature six types of inputs, DC voltage or current, potentiometer, thermocouple, AC voltage or current, RTD,and
frequency.